missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ [Arg8]-Vasopressin, MP Biomedicals™

Involved in diuresis, vasoregulation, thermoregluation and memory. [Arg8]-Vasopressin, MP Biomedicals (AVP) is an endogenous antidiuretic hormone in most mammalian species. It is released into the blood from nerve terminals in the posterior pituitary and

Brand:  MP Biomedicals™ 02152962.5

Additional Details : CAS Number : 113-79-1 Weight : 0.01000kg

Product Code. 11434091

  • 525.76 SEK / 0.50mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

AVP is implicated in a variety of physiological processes.

  • Endogenous antidiuretic hormone that regulates water balance
  • Affects rat renal medullary Na+/K+ ATPase by arginine vasopressin mediation through the V2 receptor
  • Reported to act as a neurotransmitter in the central nervous system
  • Contraction of arterioles (vasopressor action)
Specifications

Specifications

1084.239 g/mol
500 μg
RUO
95+%
[Arg8]-Vasopressin
Store at -20°C
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers