missing translation for 'onlineSavingsMsg'
Learn More

Arginase 1/ARG1/liver Arginase Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications

Brand:  Novus Biologicals NBP1-32731SS

1860.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.


 View more versions of this product

Product Code. 18515020

  • 2325.00 SEK / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Arginase 1/ARG1/liver Arginase
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
Rabbit
35 kDa
0.025 mg
Cellular Markers, Chromatin Research, Lipid and Metabolism
383
Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1:1000-1:50000, Simple Western, Flow Cytometry 5 - 10 μg/mL, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Knockdown Validated
P05089
ARG1
Arginase 1/ARG1/liver Arginase Antibody made from full length human Arginase 1 Recombinant protein.
Affinity Purified
RUO
Primary
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only