Läs mer
Arginine Vasopressin Diacetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Arginine Vasopressin Diacetate |
| CAS | 75499-44-4 |
| Molekylformel | C50H73N15O16S2 |
| Kvantitet | 5 mg |
| Synonym | 19-Amino-13-benzyl-7-(carbamoylmethyl)-4-[2-[[1-[(carbamoylmethyl)carbamoyl]-4-guanidinobutyl]carbamoyl]-1-pyrrolidinylcarbonyl]-16-p-hydroxybenzyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloeicosane-10-propionamide Diacetate, 3-(Phenylalanine)-8-arginineoxytocin, 8-L-Arginine-vasopressin Diacetate, AVP Diacetate, Arg8-vasopressin Diacetate, Arginine antidiuretic hormone Diacetate, Arginine-8-vasopressin Diacetate, Arginine-vasopressin Diacetate, Argipressin Diacetate, Leiormone Diacetate, [8-Arginine]vasopressin Diacetate, 3-(L-Phenylalanine)-8-L-arginine-oxytocin Diacetate, Pitressin Diacetate, Rindervasopressin Diacetate, L-Cysteinyl-L-tyrosyl-L-phenylalanyl-L-glutaminyl-L-asparaginyl-L-cysteinyl-L-prolyl-L-arginylglycinamide Cyclic (1→6)-Disulfide Diacetate |
| InChI formel | InChI=1S/C46H65N15O12S2.C2H4O2/c47-27-22-74-75-23-33(45(73)61-17-5-9-34(61)44(72)56-28(8-4-16-53-46(51)52)39(67)54-21-37(50)65)60-43(71)32(20-36(49)64)59-40(68)29(14-15-35(48)63)55-41(69)31(18-24-6-2-1-3-7-24)58-42(70)30(57-38(27)66)19-25-10-12-26(62)13-11-25;1-2(3)4/h1-3,6-7,10-13,27-34,62H,4-5,8-9,14-23,47H2,(H2,48,63)(H2,49,64)(H2,50,65)(H,54,67)(H,55,69)(H,56,72)(H,57,66)(H,58,70)(H,59,68)(H,60,71)(H4,51,52,53);1H3,(H,3,4)/t27-,28-,29+,30+,31-,32+,33+,34-;/m0./s1 |
| LEDER | OC(C)=O.O=C(N1CCC[C@H]1C(N[C@@H](CCCNC(N)=N)C(NCC(N)=O)=O)=O)[C@H](NC([C@@H](CC(N)=O)NC([C@@H](CCC(N)=O)NC([C@H](CC2=CC=CC=C2)N3)=O)=O)=O)CSSC[C@H](N)C(N[C@H](CC4=CC=C(O)C=C4)C3=O)=O |
| IUPAC-namn | acetic acid;(2S)-1-[(4S,7R,10R,13S,16R,19R)-19-amino-7-(2-amino-2-oxoethyl)-10-(3-amino-3-oxopropyl)-13-benzyl-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl]-N-[(2S)-1-[(2-amino-2-oxoethyl)amino]-5-(diaminomethylideneamino)-1-oxopentan-2-yl]pyrrolidine-2-carboxamide |
| Molekylvikt (g/mol) | 1204.34 |