missing translation for 'onlineSavingsMsg'
Learn More

ARHGAP25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14349-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882944

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ARHGAP25 encode negative regulators of Rho GTPases, which are implicated in actin remodeling, cell polarity, and cell migration.
Specifications

Specifications

ARHGAP25
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P42331, Q8BYW1
ARHGAP25, KAIA0053, KIAA0053
Rabbit
IgG
150 μL
-20°C
Primary
232201, 500246, 9938
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ARHGAP25
Liquid
Arhgap25
ARHGAP25 Fusion Protein Ag5691
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers