missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID4B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24499-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The exact function of C18orf25 remains unknown. There are two named isoforms.Specifications
ARID4B | |
Polyclonal | |
Unconjugated | |
ARID4B | |
ARID4B, BRCAA1, RBBP1L1, RBP1L1, SAP180 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51742 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4LE39 | |
ARID4B | |
ARID4B Fusion Protein Ag21462 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |