missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID4B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ARID4B |
---|---|
Concentration | 0.53 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822975
|
Proteintech
24499-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16812975
|
Proteintech
24499-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The exact function of C18orf25 remains unknown. There are two named isoforms.Specifications
ARID4B | |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q4LE39 | |
51742 | |
ARID4B Fusion Protein Ag21462 | |
Primary | |
-20°C | |
ARID4B |
0.53 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARID4B, BRCAA1, RBBP1L1, RBP1L1, SAP180 | |
ARID4B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |