missing translation for 'onlineSavingsMsg'
Learn More

ARIH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15006-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823974

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

E3 ubiquitin-protein ligase mediating ′Lys-48′-and ′Lys-63′-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.
Specifications

Specifications

ARIH2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95376, Q9Z1K6
ARI 2, ARI2, ariadne homolog 2 (Drosophila), ARIH2, Protein ariadne 2 homolog, TRIAD1, Triad1 protein
Rabbit
IgG
20 μL
-20°C
Primary
10425, 23807, 316005
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
Unconjugated
ARIH2
Liquid
Arih2
ARIH2 Fusion Protein Ag6507
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers