Läs mer
Aripiprazole Dimer, TRC
Produktkod. 17993405
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17993405 2.5 mg 2.5mg
17944415 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17993405 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7710252.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797986-18-5
Molekylformel C48 H56 Cl4 N6 O4
Molekylvikt (g/mol) 922.81
Synonym 7-[4-[4-[2,3-Dichloro-4-[1-[2,3-dichloro-4-[4-[4-[(2-oxo-3,4-dihydro-1H-quinolin-7-yl)oxy]butyl]piperazin-1-yl]phenyl]ethyl]phenyl]piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one, 7,7'-[Ethane-1,1-diylbis[(2,3-dichlorobenzene-4,1-diyl)piperazine-4,1-diylbutane-4,1-diyloxy]]bis[3,4.dihydroquinolin-2(1H)-one], Aripiprazole Dimer, Aripiprazole Imp. G (EP)
IUPAC-namn 7-[4-[4-[2,3-dichloro-4-[1-[2,3-dichloro-4-[4-[4-[(2-oxo-3,4-dihydro-1H-quinolin-7-yl)oxy]butyl]piperazin-1-yl]phenyl]ethyl]phenyl]piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
LEDER CC(c1ccc(N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2)c(Cl)c1Cl)c5ccc(N6CCN(CCCCOc7ccc8CCC(=O)Nc8c7)CC6)c(Cl)c5Cl
Specifikationer
Kemiskt namn eller material Aripiprazole Dimer
Kvantitet 2.5 mg
InChI formel InChI=1S/C48H56Cl4N6O4/c1-32(37-12-14-41(47(51)45(37)49)57-24-20-55(21-25-57)18-2-4-28-61-35-10-6-33-8-16-43(59)53-39(33)30-35)38-13-15-42(48(52)46(38)50)58-26-22-56(23-27-58)19-3-5-29-62-36-11-7-34-9-17-44(60)54-40(34)31-36/h6-7,10-15,30-32H,2-5,8-9,16-29H2,1H3,(H,53,59)(H,54,60)
Formel vikt 920.3117