Läs mer
Aripiprazole Hydrate, TRC
Produktkod. 30343838
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30343838 100 mg
30333778 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30343838 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA771005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Aripiprazole Hydrate
CAS 503445-20-3
Molekylformel C23 H27 Cl2 N3 O2. x[H2O]
Kvantitet 100 mg
Synonym 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone Hydrate, 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydrocarbostyril Hydrate, Abilify Hydrate, Abilitat Hydrate, OPC 14597 Hydrate, OPC 31 Hydrate, 2(1H)-Quinolinone, 7-[4-[4-(2,3-dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-, hydrate, Aripiprazole hydrate
InChI formel InChI=1S/C23H27Cl2N3O2.H2O/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18;/h3-6,8,16H,1-2,7,9-15H2,(H,26,29);1H2
LEDER O=C1NC2=CC(OCCCCN3CCN(CC3)C4=C(Cl)C(Cl)=CC=C4)=CC=C2CC1.O
IUPAC-namn 7-(4-(4-(2,3-dichlorophenyl)piperazin-1-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one hydrate
Molekylvikt (g/mol) 466.4
Formel vikt 465.16