Läs mer
Aripiprazole N,N-Dioxide, TRC
Produktkod. 17912715
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17912715 25 mg
17952255 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17912715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA77101025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 573691-13-1
Molekylformel C23 H27 Cl2 N3 O4
Molekylvikt (g/mol) 480.38
Synonym 7-[4-[4-(2,3-Dichlorophenyl)-1,4-dioxido-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone
IUPAC-namn 7-[4-[4-(2,3-dichlorophenyl)-1,4-dioxido-piperazine-1,4-diium-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
LEDER [O-][N+]1(CCCCOc2ccc3CCC(=O)Nc3c2)CC[N+]([O-])(CC1)c4cccc(Cl)c4Cl
Specifikationer
Kemiskt namn eller material Aripiprazole N,N-Dioxide
Kvantitet 25 mg
InChI formel InChI=1S/C23H27Cl2N3O4/c24-19-4-3-5-21(23(19)25)28(31)13-11-27(30,12-14-28)10-1-2-15-32-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29)
Formel vikt 497.1379