missing translation for 'onlineSavingsMsg'
Learn More

Aripiprazole N4-Oxide, TRC

Product Code. 30329869
Change view
Click to view available options
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30329869 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329869 Supplier Toronto Research Chemicals Supplier No. TRCA77102050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Aripiprazole N4-Oxide
CAS 573691-11-9
Molekylformel C23H27Cl2N3O3
Kvantitet 50 mg
Synonym 7-[4-[4-(2,3-Dichlorophenyl)-4-oxido-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone
InChI formel InChI=1S/C23H27Cl2N3O3/c24-19-4-3-5-21(23(19)25)28(30)13-11-27(12-14-28)10-1-2-15-31-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29)
LEDER [O-][N+]1(CCN(CCCCOc2ccc3CCC(=O)Nc3c2)CC1)c4cccc(Cl)c4Cl
IUPAC-namn 7-[4-[4-(2,3-dichlorophenyl)-4-oxidopiperazin-4-ium-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
Molekylvikt (g/mol) 464.38
Formel vikt 463.1429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.