Learn More
Sartorius Arium™ Advance L Filter Housing
Arium™ Advance L Filter Housing for flexible configurations of filters.
Brand: Sartorius H2O-AFHL-2
Additional Details : Weight : 1.30000kg
Description
- The Arium™ Advance L filter housing as either one-stage or two-stage allows for flexible configurations of filters for pre-treatment of water according to your system requirements.
Specifications
Double | |
Reverse Osmosis (Type 3) | |
Polypropylene Housing |
Potable Tap Water | |
Filter Housings and Holders |