Learn More
Sartorius Arium™ Advance L Filter Housing
Arium™ Advance L Filter Housing for flexible configurations of filters.
3120.00 SEK - 4710.00 SEK
Specifications
Feedwater Source | Potable Tap Water |
---|---|
Product Water Type | Reverse Osmosis (Type 3) |
Type | Filter Housings and Holders |
Material | Polypropylene Housing |
Description
- The Arium™ Advance L filter housing as either one-stage or two-stage allows for flexible configurations of filters for pre-treatment of water according to your system requirements.
Specifications
Potable Tap Water | |
Reverse Osmosis (Type 3) | |
Filter Housings and Holders | |
Polypropylene Housing |