missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16012-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the ARL (ADP-ribosylation factor-like) family of proteins, which are structurally related to ADP-ribosylation factors (ARFs). ARFs, described as activators of cholera toxin (CT) ADP-ribosyltransferase activity, regulate intracellular vesicular membrane trafficking, and stimulate a phospholipase D (PLD) isoform. Although, ARL proteins were initially thought not to activate CT or PLD, later work showed that they are weak stimulators of PLD and CT in a phospholipid dependent manner.Specifications
ARL1 | |
Polyclonal | |
Unconjugated | |
ARL1 | |
ADP ribosylation factor like 1, ARFL1, ARL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104303, 400 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P40616, P61211 | |
ARL1 | |
ARL1 Fusion Protein Ag8770 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |