Learn More
ARL2BP Mouse anti-Human, Unconjugated, Clone: 1G5C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67316-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The ARL2BP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARL2BP. This antibody recognizes human antigen. The ARL2BP antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
ARL2BP | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
1G5C2 | |
Unconjugated | |
20 μL | |
23568 | |
IgG1 |