Learn More
ARL2BP Mouse anti-Human, Unconjugated, Clone: 1G5C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67316-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16849875
  • 1656.73 SEK / 20µL
Estimated Shipment: 23-04-2024
to see stock.
Description
Description
The ARL2BP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARL2BP. This antibody recognizes human antigen. The ARL2BP antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
ARL2BP
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
1G5C2
Unconjugated
20 μL
23568
IgG1