missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL4C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10202-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport.Specifications
ARL4C | |
Polyclonal | |
Unconjugated | |
ARL4C | |
ARL4C, ARL7, LAK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10123 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P56559 | |
ARL4C | |
ARL4C Fusion Protein Ag0246 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |