Learn More
ARMC1 Recombinant Protein, Abnova
Brand:  Abnova H00055156-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16131203
  • 5030.41 SEK / 25µg
Estimated Shipment: 18-11-2021
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH11607
Solution
55156
ARMC1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Arcp/FLJ10511
ARMC1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
57.1
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARMC1