missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMC5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24833-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
ARMC5 | |
Polyclonal | |
Unconjugated | |
ARMC5 | |
armadillo repeat containing 5, ARMC5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233912, 79798 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5EBP3, Q96C12 | |
ARMC5 | |
ARMC5 Fusion Protein Ag21171 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |