missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARMCX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12200-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The armadillo (ARM) repeat family of proteins are related to the Drosophila melanogaster armadillo protein, a protein essential for wingless signal transduction. ARM proteins are involved in a variety of processes such as cell migration, cell proliferation, tissue maintece and tumorigenesis, and they also function in signal transduction and the maintece of overall cell structure. ARMCX2 (armadillo repeat containing, X-linked 2), also known as ALEX2, is a 632 amino acid single-pass membrane protein that contains three ARM repeats and is highly expressed in testis, brain, ovary, heart, colon, spleen and prostate, where it is thought to play a role in tumor suppression. The gene encoding ARMCX2 maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1, 000 genes.Specifications
ARMCX2 | |
Polyclonal | |
Unconjugated | |
ARMCX2 | |
ALEX2, ARMCX2, KIAA0512, Protein ALEX2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
367903, 67416, 9823 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6A058, Q7L311 | |
Armcx2 | |
ARMCX2 Fusion Protein Ag2839 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |