missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ARNTL (Human) Recombinant Protein
Human ARNTL full-length ORF ( AAH41129, 1 a.a. - 625 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000406-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MADQRMDISSTISDFMSPGPTDLLSSSLGTSGVDCNRKRKGSSTDYQESMDTDKDDPHGRLEYTEHQGRIKNAREAHSQIEKRRRDKMNSFIDELASLVPTCNAMSRKLDKLTVLRMAVQHMKTLRGATNPYTEANYKPTFLSDDELKHLILRAADGFLFVVGCDRGKILFVSESVFKILNYSQNDLIGQSLFDYLHPKDIAKVKEQLSSSDTAPRERLIDAKTGLPVKTDITPGPSRLCSGARRSFFCRMKCNRPSVKVEDKDFPSTCSKKKDRKSFCTIHSTGYLKSWPPTKMGLDEDNEPDNEGCNLSCLVAIGRLHSHVVPQPVNGEIRVKSMEYVSRHAIDGKFVFVDQRATAILAYLPQELLGTSCYEYFHQDDIGHLAECHRQVLQTREKITTNCYKFKIKDGSFITLRSRWFSFMNPWTKEVEYIVSTNTVVLANVLEGGDPTFPQLTASPHSMDSMLPSGEGGPKRTHPTVPGIPGGTRAGAGKIGRMIAEEIMEIHRIRGSSPSSCGSSPLNITSTPPPDASSPGGKKILNGGTPDIPSSGLLSGQAQENPGYPYSDSSSILGENPHIGIDMIDNDQGSSSPSNDEAAMAVIMSLLEADAGLGGPVDFSDLPWPL
Specifications
AAH41129 | |
aryl hydrocarbon receptor nuclear translocator-like | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARNTL | |
GST |
406 | |
Wheat germ expression system | |
10 μg | |
BMAL1, BMAL1c, JAP3, MGC47515, MOP3, PASD3, TIC, bHLHe5 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |