missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10930-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3 , which results in the production of a fusion protein.Specifications
ARPC4 | |
Polyclonal | |
Unconjugated | |
ARPC4 | |
ARC20, Arp2/3 complex 20 kDa subunit, ARPC4, p20 Arc | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10093, 297518, 68089 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P59998, P59999 | |
Arpc4 | |
ARPC4 Fusion Protein Ag1366 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |