Läs mer
Arprinocid, TRC
Produktkod. 17931295
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17931295 100 mg
17963105 250 mg
17994535 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17931295 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA735500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55779-18-5
Molekylformel C12 H9 Cl F N5
Molekylvikt (g/mol) 277.68
Synonym 9-[(2-Chloro-6-fluorophenyl)methyl]-9H-purin-6-amine, 6-Amino-9-(2-chloro-6-fluorobenzyl)purine, Arpocox, Arprinocid, MK 302
IUPAC-namn 9-[(2-chloro-6-fluorophenyl)methyl]purin-6-amine
LEDER Nc1ncnc2c1ncn2Cc3c(F)cccc3Cl
Specifikationer
Kemiskt namn eller material Arprinocid
Kvantitet 100 mg
InChI formel InChI=1S/C12H9ClFN5/c13-8-2-1-3-9(14)7(8)4-19-6-18-10-11(15)16-5-17-12(10)19/h1-3,5-6H,4H2,(H2,15,16,17)
Formel vikt 277.0531