missing translation for 'onlineSavingsMsg'
Learn More

ARSA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19061-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878844

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy , a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
Specifications

Specifications

ARSA
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15289, P50428
ARSA, arylsulfatase A, ASA, Cerebroside sulfatase, MLD
Rabbit
IgG
20 μL
-20°C
Primary
11883, 315222, 410
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARSA
Liquid
Arsa
ARSA Fusion Protein Ag13569
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers