Läs mer
Arsenic (III) Standard: As+3 @ 100 μg/mL in 2% HCl, VHG™
Produktkod. 17940536
Change view
Kvantitet:
50 ml
100 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17940536 100 ml
17927819 50 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17940536 Leverantör VHG Labs Leverantörsnummer VHGSPAS3100
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Arsenic (III) Standard: As+3 @ 100 μg/mL in 2% HCl
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Arsenic+3
CAS 22541-54-4
Metoder Speciation
Analyt- eller komponentnamn Arsenic+3
Frakt skick Room Temperature
Molekylformel As
Synonym As3+
InChI formel InChI=1S/As/q+3
LEDER [As+3]
IUPAC-namn arsenic(3+)
Lösningsmedel eller matris 2% HCl
Molekylvikt (g/mol) 74.922
Kvantitet 100 ml
Formel vikt 74.922 g/mol