Läs mer
Arsenic Oxide (As2O5) Hydrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Arsenic Oxide (As2O5) Hydrate |
| CAS | 2179255-37-7 |
| Molekylformel | As2O5 xH2O |
| Kvantitet | 1 g |
| Synonym | Arsenic (V) oxide Hydrate, Arsenic anhydride Hydrate, Arsenic oxide (As4O10) Hydrate, Arsenic pentoxide Hydrate, Arsenic(V) pentoxide Hydrate, Diarsenic pentoxide Hydrate, Diarsonic pentoxide Hydrate |
| InChI formel | InChI=1S/As2O5/c3-1(4)2(5,6)7 |
| LEDER | [O][As](=O)([O])[As](=O)=O |
| IUPAC-namn | arsoarsonic acid |
| Molekylvikt (g/mol) | 231.86 |
| Formel vikt | 229.8178 |