Läs mer
Arsenobetaine Bromide, TRC
Produktkod. 30348024
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30348024 250 mg
1 options
Produktkod. 30348024 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA778500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Arsenobetaine bromide
CAS 71642-15-4
Anteckningar om renhetsgrad HPLC
Molekylformel C5 H12 As O2 . Br
Kvantitet 250 mg
Synonym (Carboxymethyl)trimethylarsonium Bromide, NSC 274266
InChI formel InChI=1S/C5H11AsO2.BrH/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1H
LEDER [Br-].C[As+](C)(C)CC(=O)O
IUPAC-namn carboxymethyl(trimethyl)arsanium;bromide
Molekylvikt (g/mol) 258.97
Formel vikt 257.9237