missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ART3 (Human) Recombinant Protein
Human ART3 full-length ORF ( AAH08397, 27 a.a. - 389 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000419-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: EVLDMADNAFDDEYLKCTDRMEIKYVPQLLKEEKASHQQLDTVWENAKAKWAARKTQIFLPMNFKDNHGIALMAYISEAQEQTPFYHLFSEAVKMAGQSREDYIYGFQFKAFHFYLTRALQLLRKPCEASSKTVVYRTSQGTSFTFGGLNQARFGHFTLAYSAKPQAANDQLTVLSIYTCLGVDIENFLDKESERITLIPLNEVFQVSQEGAGNNLILQSINKTCSHYECAFLGGLKTENCIENLEYFQPIYVYNPGEKNQKLEDHSEKNWKLEDHGEKNQKLEDHGVKILEPTQIPGMKIPEPFPLPEDKSQGNINNPTPGPVPVPGPKSHPSASLGKLLLPQFGMVIILISVSAINLFVAL
Specifications
AAH08397 | |
ADP-ribosyltransferase 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ART3 | |
GST |
419 | |
Wheat germ expression system | |
25 μg | |
FLJ26404 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |