missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ART5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550335
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The antibody detects endogenous levels of total ART5 protein.
The protein encoded by this gene belongs to the ARG-specific ADP-ribosyltransferase family. Proteins in this family regulate the function of target proteins by attaching ADP-ribose to specific amino acid residues in their target proteins. The mouse homolog lacks a glycosylphosphatidylinositol-anchor signal sequence and is predicted to be a secretory enzyme. Transcript variants with different 5' UTRs, but encoding the same protein have been found for this gene.
Specifikationer
| ART5 | |
| Polyclonal | |
| Unconjugated | |
| ART5 | |
| ADP-ribosyltransferase 5; ADP-ribosyltransferase C2 and C3 toxin-like 5; Art5; ARTC5; Ecto-ADP-ribosyltransferase 5; mono(ADP-ribosyl)transferase 5; NAD(P)(+)--arginine ADP-ribosyltransferase 5; UNQ575/PRO1137; YAC-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116969 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96L15 | |
| ART5 | |
| Fusion protein corresponding to a region derived from internal residues of human ADP-ribosyltransferase 5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering