missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11354-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody detects the 43kD ART5 protein in mouse liver and testis.
The protein encoded by this gene belongs to the ARG-specific ADP-ribosyltransferase family. Proteins in this family regulate the function of target proteins by attaching ADP-ribose to specific amino acid residues in their target proteins. The mouse homolog lacks a glycosylphosphatidylinositol-anchor signal sequence and is predicted to be a secretory enzyme. Several transcripts encoding different isoforms have been found for this gene.Specifications
ART5 | |
Polyclonal | |
Unconjugated | |
ART5 | |
ADP ribosyltransferase 5, ART5, Ecto ADP ribosyltransferase 5, Mono(ADP ribosyl)transferase 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116969, 11875, 259167 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70352, Q5XHY4, Q96L15 | |
Art5 | |
ART5 Fusion Protein Ag1786 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |