Läs mer
Artemisinic Aldehyde, TRC
Produktkod. 30137304
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137304 25 mg -
30006040 2.5 mg 2.5mg
30003333 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137304 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA77751025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 125276-60-0
Molekylformel C15 H22 O
Molekylvikt (g/mol) 218.33
Synonym 1-Naphthaleneacetaldehyde, 1,2,3,4,4a,5,6,8a-octahydro-4,7-dimethyl-α-methylene-, (1R,4R,4aS,8aR)-, 1-Naphthaleneacetaldehyde, 1,2,3,4,4a,5,6,8a-octahydro-4,7-dimethyl-α-methylene-, [1R-(1α,4β,4aβ,8aβ)]-, (1R,4R,4aS,8aR)-1,2,3,4,4a,5,6,8a-Octahydro-4,7-dimethyl-α-methylene-1-naphthaleneacetaldehyde, Artemisinic aldehyde
IUPAC-namn 2-[(1R,4R,4aS,8aR)-4,7-dimethyl-1,2,3,4,4a,5,6,8a-octahydronaphthalen-1-yl]prop-2-enal
LEDER C[C@@H]1CC[C@H]([C@H]2C=C(C)CC[C@@H]12)C(=C)C=O
Specifikationer
Kemiskt namn eller material Artemisinic Aldehyde
Analyt- eller komponentnamn Artemisinic Aldehyde
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C15H22O/c1-10-4-6-13-11(2)5-7-14(12(3)9-16)15(13)8-10/h8-9,11,13-15H,3-7H2,1-2H3/t11-,13+,14+,15+/m1/s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 25 mg
Formel vikt 218.1671
Procent renhet >95%