missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Arterolane, MedChemExpress
Description
Arterolane is an antimalarial agent, with IC50 of both 1.1 nM against P. falciparum Ro73 and W2, respectively.
Specifications
Specifications
| Kemiskt namn eller material | Arterolane |
| Fysisk form | Solid |
| CAS | 664338-39-0 |
| Färg | White |
| Kvantitet | 5 mg |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C22H36N2O4 |
| För användning med (applikation) | COVID-19-immunoregulation |
| Synonym | OZ 277 RBx 11160 |
| Löslighetsinformation | DMSO : 100 mg/mL (254.76 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?