Läs mer
Articaine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Articaine hydrochloride |
| CAS | 23964-57-0 |
| Molekylformel | C13 H20 N2 O3 S . Cl H |
| Kvantitet | 10 mg |
| Synonym | Articaine Hydrochloride, 2-Thiophenecarboxylic acid, 4-methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-, methyl ester, hydrochloride (1:1), 2-Thiophenecarboxylic acid, 4-methyl-3-[2-(propylamino)propionamido]-, methyl ester, monohydrochloride (8CI), 2-Thiophenecarboxylic acid, 4-methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-, methyl ester, monohydrochloride (9CI), Carticain, Carticaine chlorhydrate, Carticaine hydrochloride, HOE 045, HOE 40045, Methyl 4-methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylate hydrochloride, Septanest, Ubistesine, Ultacain, Ultracain, Ultracaine |
| InChI formel | InChI=1S/C13H20N2O3S.ClH/c1-5-6-14-9(3)12(16)15-10-8(2)7-19-11(10)13(17)18-4;/h7,9,14H,5-6H2,1-4H3,(H,15,16);1H |
| LEDER | Cl.CCCNC(C)C(=O)Nc1c(C)csc1C(=O)OC |
| IUPAC-namn | methyl 4-methyl-3-[2-(propylamino)propanoylamino]thiophene-2-carboxylate;hydrochloride |
| Molekylvikt (g/mol) | 320.84 |
| Formel vikt | 320.0961 |