missing translation for 'onlineSavingsMsg'
Läs mer

Articaine, TRC

Product Code. 30348368
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30348368 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348368 Supplier Toronto Research Chemicals Supplier No. TRCA777900250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Articaine
CAS 23964-58-1
Molekylformel C13 H20 N2 O3 S
Kvantitet 250 mg
Synonym 4-Methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylic Acid Methyl Ester, 4-Methyl-3-[2-(propylamino)propionamido]-2-thiophenecarboxylic Acid Methyl Ester (8CI), Articaine, Carticaine
InChI formel InChI=1S/C13H20N2O3S/c1-5-6-14-9(3)12(16)15-10-8(2)7-19-11(10)13(17)18-4/h7,9,14H,5-6H2,1-4H3,(H,15,16)
LEDER CCCNC(C)C(=O)Nc1c(C)csc1C(=O)OC
IUPAC-namn methyl 4-methyl-3-[2-(propylamino)propanoylamino]thiophene-2-carboxylate
Molekylvikt (g/mol) 284.37
Formel vikt 284.1195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.