Läs mer
ARV-825, MedChemExpress
Produktkod. 16477218
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16477218 10 mg 10mg
16459747 5 mg -
16473237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16477218 Leverantör Medchem Express Leverantörsnummer HY16954
MedChemExpress ARV-825 is a PROTAC connected by ligands for Cereblon and BRD4. ARV-825 binds to BD1 and BD2 of BRD4 with Kds of 90 and 28 nM, respectively.
ARV-825 is a PROTAC connected by ligands for Cereblon and BRD4. ARV-825 binds to BD1 and BD2 of BRD4 with Kds of 90 and 28 nM, respectively.
Specifikationer
Kemiskt namn eller material ARV-825
CAS 1818885-28-7
Färg Deep Yellow
Anteckningar om renhetsgrad Research
Molekylformel C46H47ClN8O9S
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 50 mg/mL (54.15 mM)
LEDER CC1=NN=C2N1C(SC(C)=C3C)=C3C(C4=CC=C(Cl)C=C4)=N[C@H]2CC(NC5=CC=C(OCCOCCOCCOCCNC6=C(C(N(C7CCC(NC7=O)=O)C8=O)=O)C8=CC=C6)C=C5)=O
Molekylvikt (g/mol) 923.43
Formel vikt 923.43
Procent renhet 98.01%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death