missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARX Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20739-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to ARX. This is a rabbit polyclonal antibody raised against a peptide mapping within human ARX.
Arx contains two conserved domains, a C-peptide (or aristaless domain) and the prd-like class homeobox domain. It is a member of the group-II aristaless-related protein family whose members are expressed primarily in the central and/or peripheral nervous system. This protein is thought to be involved in CNS development. Mutations in the Arx gene cause X-linked mental retardation and epilepsy.Specifications
ARX | |
Polyclonal | |
Unconjugated | |
ARX | |
aristaless related homeobox, ARX, Homeobox protein ARX, ISSX, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11878, 170302 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35085, Q96QS3 | |
ARX | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |