Läs mer
Arylex, TRC
Produktkod. 30344823
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30344823 10 mg
1 options
Produktkod. 30344823 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79489010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Halauxifen-methyl
CAS 943831-98-9
Molekylformel C14 H11 Cl2 F N2 O3
Kvantitet 10 mg
Synonym Arylex, Arylex active, Halauxifen-methyl, XDE 729
InChI formel InChI=1S/C14H11Cl2FN2O3/c1-21-13-7(15)4-3-6(11(13)17)9-5-8(18)10(16)12(19-9)14(20)22-2/h3-5H,1-2H3,(H2,18,19)
LEDER COC(=O)c1nc(cc(N)c1Cl)c2ccc(Cl)c(OC)c2F
IUPAC-namn methyl 4-amino-3-chloro-6-(4-chloro-2-fluoro-3-methoxyphenyl)pyridine-2-carboxylate
Molekylvikt (g/mol) 345.15
Formel vikt 344.0131