missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Arylsulfatase G Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597473
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Arylsulfatase G Polyclonal Antibody for Western Blot, IHC (P), ELISA
Sulfatases, such as ARSG, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules. This gene, a novel arylsulfatase, has activity toward pseudosubstrates including p-nitrocatechol sulfate and 4-methylumbelliferyl sulfate. Activity is competitively inhibited by phosphate. The unprocessed protein is active as a 63-kDa monomer and demonstrates an acidic pH optimum as typically seen for lysosomal sulfatases. The protein accumulates within lysosomes and is also a glycoprotein that binds specifically to mannose 6-phosphate receptors.
Specifikationer
| Arylsulfatase G | |
| Polyclonal | |
| Unconjugated | |
| ARSG | |
| 6330406P08Rik; AI846872; Arsg; Arylsulfatase G; ASG; KIAA1001; RGD1306571; UNQ839/PRO1777 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 22901 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96EG1 | |
| ARSG | |
| Recombinant Human Arylsulfatase G protein (17-525AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering