Läs mer
Tocris Bioscience™ AS 2444697
Beskrivning
AS 2444697 is a potent and selective interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor (IC50 = 21 nM). Displays 30-fold selectivity for IRAK4 over IRAK1. Inhibits LPS-induced TNF-α and IL-6 production in PBMCs in vitro. Renoprotective and anti-inflammatory in a rodent model of chronic kidney disease.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-[3-Aminocarbonyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-yl]-2-(2-methyl-4-pyridinyl)-4-oxazolecarboxamide hydrochloride |
| CAS | 1287665-60-4 |
| Kvantitet | 50 mg |
| Mål | Other Kinase Inhibitors |
| Molekylformel | C19H20N6O4.HCl |
| Formel vikt | 432.86 |
| Renhet | >98% |