missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASBT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ASBT |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823825
|
Proteintech
25245-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813825
|
Proteintech
25245-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ASBT | |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P70172, Q12908 | |
20494, 6555 | |
ASBT Fusion Protein Ag18808 | |
Primary | |
-20°C | |
SLC10A2 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ASBT, IBAT, ISBT, NTCP2, SLC10A2 | |
SLC10A2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |