Läs mer
Asciminib, MedChemExpress
Produktkod. 16424956
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16424956 100 mg
16423928 10 mg
16416647 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16424956 Leverantör Medchem Express Leverantörsnummer HY104010
MedChemExpress Asciminib (ABL001) is a potent and selective allosteric BCR-ABL1 inhibitor, which inhibits Ba/F3 cells grown with an IC50 of 0.25 nM.
Asciminib (ABL001) is a potent and selective allosteric BCR-ABL1 inhibitor, which inhibits Ba/F3 cells grown with an IC50 of 0.25 nM.
Specifikationer
Kemiskt namn eller material Asciminib
CAS 1492952-76-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C20H18ClF2N5O3
Kvantitet 100 mg
Synonym ABL001
Löslighetsinformation DMSO : ≥ 100 mg/mL (222.30 mM)
LEDER O=C(C1=CC(C2=NNC=C2)=C(N3C[C@H](O)CC3)N=C1)NC4=CC=C(OC(F)(Cl)F)C=C4
Molekylvikt (g/mol) 449.84
Formel vikt 449.84
Procent renhet 99.78%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease