Learn More
Invitrogen™ ASHG/Fetuin A Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHAHSGX5
Additional Details : Weight : 0.33820kg
Description
The Human ASHG/ Fetuin A ELISA quantitates Hu ASHG in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ASHG. Principle of the method The Human ASHG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.Specifications
P02765 | |
0.2 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
197 | |
<12% | |
Chromogen, Adhesive Plate Covers, Assay Diluent Concentrate, Stop Solution, SAV-HRP, Wash Buffer, Pre-coated 96 well plates, Biotinylated Detection Antibody, Standard | |
5 x 96 Tests | |
Inflammation | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
0.205 ng/mL to 50 ng/mL | |
0.2 ng/mL to 50 ng/mL | |
Biotin | |
Kinase & Inhibitors | |
This ELISA antibody pair detects human Fetuin A. Other species not determined. | |
Human | |
Colorimetric Microplate Reader | |
A2HS,AHS,FETUA,HSGA | |
<10% | |
HRP | |
RUO | |
Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L | |
PRO2743, A2HS, AHS, FETUA, HSGA | |
1 hr. 20 min. |