missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ASL (Human) Recombinant Protein

Human ASL full-length ORF ( AAH08195, 1 a.a. - 464 a.a.) recombinant protein with GST-tag at N-terminal.

4515.00 SEK - 6845.00 SEK

Specifications

Accession Number AAH08195
Gene ID (Entrez) 435
Name argininosuccinate lyase
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16151941
View Documents Promotion Details
Abnova™
H00000435-P01.10UG
10 μg
4515.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16161941
View Documents Promotion Details
Abnova™
H00000435-P01.25UG
25 μg
6845.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sequence: MASESGKLWGGRFVGAVDPIMEKFNASIAYDRHLWEVDVQGSKAYSRGLEKAGLLTKAEMDQILHGLDKVAEEWAQGTFKLNSNDEDIHTANERRLKELIGATAGKLHTGRSRNDQVVTDLRLWMRQTCSTLSGLLWELIRTMVDRAEAERDVLFPGYTHLQRAQPIRWSHWILSHAVALTRDSERLLEVRKRINVLPLGSGAIAGNPLGVDRELLRAELNFGAITLNSMDATSERDFVAELLFWASLCMTHLSRMAEDLILYCTKEFSFVQPSDAYSTGSSLMPQKKNPDSLELIRSKAGRVFGRCAGLLMTLKGLPSTYNKDLQEDKEAVFEVSDTMSAVLQVATGVISTLQIHQENMGQALSPDMLATDLAYYLVRKGMPFRQAHEASGKAVFMAETKGVALNQLSLQELQTISPLFSGDVICVWDYGHSVEQYGALGGTARSSVDWQIRQVRALLQAQQA
Specifications

Specifications

AAH08195
argininosuccinate lyase
125% SDS-PAGE Stained with Coomassie Blue
ASAL
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
435
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASL
GST
Videos
SDS
Documents

Documents

Product Certifications