Läs mer
ASM-DNT, TRC
Produktkod. 30136593
Change view
Kvantitet:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136593 2.5 g 2.5g
30136269 500 mg -
30010225 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136593 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4934302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93-46-9
Molekylformel C26 H20 N2
Molekylvikt (g/mol) 360.4504
Synonym 1,4-Benzenediamine, N,N'-di-2-naphthalenyl- (9CI), p-Phenylenediamine, N,N'-di-2-naphthyl- (6CI,8CI), N1,N4-Di-2-naphthalenyl-1,4-benzenediamine, 1,4-Bis(2-naphthylamino)benzene, ASM-DNT, Aceto DIPP, AgeRite W, AgeRite White, Anchor DNPD, Antage F, Antigene F, Antioxidant 123, Antioxidant DNP, DNP, DNPD, Diafen NN, N,N'-Bis(2-naphthyl)-1,4-phenylenediamine, N,N'-Bis(2-naphthyl)-p-phenylenediamine, N,N'-Bis(β-naphthyl)-p-phenylenediamine, N,N'-Di(2-naphthyl)-p-phenylenediamine, N,N'-Di-2-naphthyl-1,4-benzenediamine, N,N'-Di-2-naphthyl-1,4-phenylenediamine, N,N'-Di-β-naphthyl-p-phenylenediamine, N,N'-di-2-Naphthyl-p-phenyldiamine, N,N'-di-β-Naphthyl-p-phenylenediamine, N,N'-p-Phenylenebis[2-naphthylamine], NSC 3410, Nocrac White, Nonflex F, Nonox CI, Ouchi Shinko White F, Santowhite CI, Tisperse MB 2X, s-Di-β-Naphthyl-p-phenylenediamine, sym-Di-β-naphthyl-p-phenylenediamine, N,N'-Bis-(2-naphthyl)-p-phenylenediamine (DNPD)
IUPAC-namn 1-N,4-N-dinaphthalen-2-ylbenzene-1,4-diamine
LEDER N(c1ccc(Nc2ccc3ccccc3c2)cc1)c4ccc5ccccc5c4
Specifikationer
Kemiskt namn eller material N,N'-Bis-(2-naphthyl)-p-phenylenediamine (DNPD)
Kvantitet 2.5 g
InChI formel InChI=1S/C26H20N2/c1-3-7-21-17-25(11-9-19(21)5-1)27-23-13-15-24(16-14-23)28-26-12-10-20-6-2-4-8-22(20)18-26/h1-18,27-28H
Formel vikt 360.1626