missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15450-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.Specifications
ASNA1 | |
Polyclonal | |
Unconjugated | |
ASNA1 | |
ARSA, ARSA I, ARSA1, ASNA1, ATPase ASNA1, hARSA I, hASNA I, TRC40 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288919, 439, 56495 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43681, O54984 | |
Asna1 | |
ASNA1 Fusion Protein Ag7713 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |