Learn More
Abcam Asparagine Assay Kit (Fluorometric)
Product Code. 32120195 Shop All Abcam Products
Change view
Click to view available options
100 Tests
Product Code.
32120195 100 Tests
1 options
Product Code. 32120195 Supplier Abcam Supplier No. ab273333100Test
 Restricted Item
Beräknad leverans: 25-11-2026
Measure Asparagine in Cell Samples, Cell Culture Media, Biofluids with Asparagine Assay Kit AB273333. Cited in 2 publications. Use with Microplate.
Measure Asparagine in Cell Samples, Cell Culture Media, Biofluids with Asparagine Assay Kit AB273333. Cited in 2 publications. Use with Microplate.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO