missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Aspon 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | Aspon |
| CAS | 3244-90-4 |
| Analyt- eller komponentnamn | Aspon |
| Frakt skick | Room Temperature |
| Molekylformel | C12 H28 O5 P2 S2 |
| Koncentration | 100 μg/mL |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Propyl thiopyrophosphate ([(PrO)2P(S)]2O) (6CI,7CI), Thiodiphosphoric acid ([(HO)2P(S)]2O), tetrapropyl ester (9CI), Thiopyrophosphoric acid ([(HO)2PS]2O), tetrapropyl ester (8CI), A 42, ASP 51, Aspon, E 8573, NPD, NSC 2550, O,O,O,O-Tetrapropyl dithiopyrophosphate, Tetra-n-propyl dithionopyrophosphate, Tetra-n-propyl dithiopyrophosphate |
| InChI formel | InChI=1S/C12H28O5P2S2/c1-5-9-13-18(20,14-10-6-2)17-19(21,15-11-7-3)16-12-8-4/h5-12H2,1-4H3 |
| LEDER | CCCOP(=S)(OCCC)OP(=S)(OCCC)OCCC |
| Visa mer |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?