Läs mer
Astemizole-d3, TRC
Produktkod. 30336519
Change view
Click to view available options
Kvantitet:
1 mg
Product Code. Kvantitet
30336519 1 mg
1 options
Product Code. 30336519 Supplier Toronto Research Chemicals Supplier No. TRCA7900321MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Astemizole-d3
CAS 1189961-39-4
Molekylformel C28H28D3FN4O
Kvantitet 1 mg
Synonym Astemizole D3
InChI formel InChI=1S/C28H31FN4O/c1-34-25-12-8-21(9-13-25)14-17-32-18-15-24(16-19-32)30-28-31-26-4-2-3-5-27(26)33(28)20-22-6-10-23(29)11-7-22/h2-13,24H,14-20H2,1H3,(H,30,31)/i1D3
LEDER [2H]C([2H])([2H])Oc1ccc(CCN2CCC(CC2)Nc3nc4ccccc4n3Cc5ccc(F)cc5)cc1
IUPAC-namn 1-[(4-fluorophenyl)methyl]-N-[1-[2-[4-(trideuteriomethoxy)phenyl]ethyl]piperidin-4-yl]benzimidazol-2-amine
Molekylvikt (g/mol) 461.59