missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ Astemizole
Description
Orally active, potent histamine H1 antagonist (IC50 = 4 nM) that displays 20-fold, > 250-fold and > 250-fold selectivity over 5-HT, dopamine and muscarinic acetylcholine receptors respectively. Exhibits antimalarial activity in multidrug resistant strains in vitro (IC50 = 227 - 734 nM). Also potent KV11.1 (hERG) channel blocker (IC50 = 0.9 nM) that displays cardiotoxicity in vivo.
Specifications
Specifications
| Kemiskt namn eller material | Astemizole |
| CAS | 68844-77-9 |
| Kvantitet | 50 mg |
| Molekylformel | C28H31FN4O |
| Formel vikt | 458.57 |
| Renhet | >99% |
| Molekylvikt (g/mol) | 458.581 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?