Läs mer
Astemizole, TRC
Produktkod. 17920935
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17920935 25 mg
17941515 50 mg
17934865 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17920935 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79003025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68844-77-9
Molekylformel C28 H31 F N4 O
Molekylvikt (g/mol) 458.57
Synonym 1H-Benzimidazol-2-amine, 1-[(4-fluorophenyl)methyl]-N-[1-[2-(4-methoxyphenyl)ethyl]-4-piperidinyl]-, Astemisan, Astemizole, Hismanal, Histamen, Histaminos, Histazol, Kelp, Laridal, Metodik, NSC 329963, Novo-Nastizol A, Paralergin, R 42512, R 43512, Retolen, Waruzol
IUPAC-namn 1-[(4-fluorophenyl)methyl]-N-[1-[2-(4-methoxyphenyl)ethyl]piperidin-4-yl]benzimidazol-2-amine
LEDER COc1ccc(CCN2CCC(CC2)Nc3nc4ccccc4n3Cc5ccc(F)cc5)cc1
Specifikationer
Kemiskt namn eller material Astemizole
Analyt- eller komponentnamn Astemizole
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C28H31FN4O/c1-34-25-12-8-21(9-13-25)14-17-32-18-15-24(16-19-32)30-28-31-26-4-2-3-5-27(26)33(28)20-22-6-10-23(29)11-7-22/h2-13,24H,14-20H2,1H3,(H,30,31)
Lösningstyp Neat
Industry Type Forensics
Kvantitet 25 mg
Formel vikt 458.2482
Procent renhet >95%